Check Order Status
1,5-DipHenylcarbazone, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
1,5-DipHenylcarbazone, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCD485045
also known as FGCD/4850/45
10 g
Glass Jar
Catalog number
FGCD485045
also known as FGCD/4850/45
Price (MYR)
-
Quantity:
10 g
Packaging:
Glass Jar
Request bulk or custom format
Specifications
CAS
538-62-5
Melting Point
153°C
Color
Orange
Packaging
Glass Jar
Molecular Formula
C13H12N4O
View more