Check Order Status
1,4-Dioxane, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™
Element Search
Structure Search
1,4-Dioxane, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCD450008
also known as FGCD/4500/08
500 mL
Amber Glass Bottle
Catalog number
FGCD450008
also known as FGCD/4500/08
Price (MYR)
328.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
328.00
EA
Add to cart
Specifications
pH
6 to 8
Melting Point
12°C
CAS
123-91-1
Color
Colorless
Boiling Point
101°C
View more