Check Order Status
Dichloroacetic Acid, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Dichloroacetic Acid, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCD1550PB15
also known as FGCD/1550/PB15
1 L
Catalog number
FGCD1550PB15
also known as FGCD/1550/PB15
Price (MYR)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS
79-43-6
Melting Point
9°C
Color
Yellow
pH
1.2
Boiling Point
194°C
View more