Check Order Status
1,2-Diaminoethane, Anhydrous, SLR, Fisher Chemical
Element Search
Structure Search
1,2-Diaminoethane, Anhydrous, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCD0370PB17
also known as FGCD/0370/PB17
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
107-15-3
Melting Point
11°C
Color
Colorless
pH
12.2
Boiling Point
117°C
View more