Check Order Status
Citric Acid Monohydrate, Extra Pure, SLR, Granular, Fisher Chemical™
Element Search
Structure Search
Citric Acid Monohydrate, Extra Pure, SLR, Granular, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 kg
500 g
1 kg
Packaging:
Plastic powder jar
Catalog number
FGCC616062
also known as FGCC/6160/62
Price (MYR)
410.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
410.00
EA
Add to cart
Specifications
pH
1.8
CAS
5949-29-1
Melting Point
135°C
Color
White
Molecular Formula
C6H10O8
View more