Check Order Status
Carborundum, Technical, SLR, Powder, Medium, about 165μm (80 Grit), Fisher Chemical
Element Search
Structure Search
Carborundum, Technical, SLR, Powder, Medium, about 165μm (80 Grit), Fisher Chemical
Have Questions?
Catalog number
FGCC309153
also known as FGCC/3091/53
Price (MYR)
-
Request bulk or custom format
Chemical Identifiers
CAS
409-21-2
Specifications
Color
Black
Physical Form
Solid
Packaging
Plastic Powder Jar
CAS
409-21-2
Melting Point
2700°C
View more