Check Order Status
Caffeine, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Caffeine, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCC080048
also known as FGCC/0800/48
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
58-08-2
Melting Point
234°C
Color
White
pH
5.5 to 6.5
Packaging
Plastic powder jar
View more