Check Order Status
Cadmium, Technical, Metal, Granulated, SLR, Fisher Chemical
Element Search
Structure Search
Cadmium, Technical, Metal, Granulated, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCC008048
also known as FGCC/0080/48
Price (MYR)
-
Request bulk or custom format
Specifications
Color
Silver
CAS
7440-43-9
Melting Point
321°C
Boiling Point
765°C
Quantity
100 g
View more