Adenosine 5'-triphosphate Disodium Salt Trihydrate
Adenosine 5'-triphosphate Disodium Salt Trihydrate
Actual product may vary
Adenosine 5'-triphosphate Disodium Salt Trihydrate
Adenosine 5'-triphosphate Disodium Salt Trihydrate
Fisher Chemical
Adenosine 5'-triphosphate Disodium Salt Trihydrate
Have Questions?
Catalog NumberQuantity
FGCBP413-2525 g
Catalog number FGCBP413-25
Price (MYR)
946.00
EA
Add to cart
Quantity:
25 g
Price (MYR)
946.00
EA
Add to cart
Disodium Salt ATP
  • C10H14N5O13P3Na2·3H2O; F.W. 605.24
  • CAS: 51963-61-2
  • Molar Extinction Coefficient (259nm, phosphate buffer, pH 7.0); >15,000
  • Solubility (5% in H2O, 25°C): clear and haze-free
  • Lead: ≤0.002%
Suitable for use in many molecular biology applications
Specifications
FormatSolid
pH7
Molecular Weight (g/mol)605.24
Quantity25 g
ColorWhite
For Use With (Application)Molecular biology
Unit SizeEA
Contents & Storage
<0°C