Check Order Status
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number
FGCBP332-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Phosphate Dibasic Anhydrous
CAS
7558-79-4
ChemAlert Storage Symbol
Gray
Color
White
Health Hazard 1
Signal Word: CAUTION!
View more
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)
This buffer component is commonly used in tissue culture and molecular biology applications.