Check Order Status
Pyridoxine Hydrochloride, Fisher BioReagents
Element Search
Structure Search
Pyridoxine Hydrochloride, Fisher BioReagents
Have Questions?
Change view
Quantity:
10 g
50 g
Packaging:
Amber Glass
Catalog number
FGCBP2677-10
Price (MYR)
-
Quantity:
10 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Chemical Name or Material
Pyridoxine Hydrochloride
CAS
58-56-0
ChemAlert Storage Symbol
Gray
Melting Point
214°C
Color
White
View more
Pyridoxine HCl is used as a vitamin (enzyme cofactor).