Check Order Status
TE Buffer, Tris-EDTA, 1X Solution, pH 8.0, Molecular Biology, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
TE Buffer, Tris-EDTA, 1X Solution, pH 8.0, Molecular Biology, Fisher BioReagents
Have Questions?
Change view
Quantity:
1 L
100 mL
500 mL
Packaging:
View all
Nalgene Poly Bottle
Catalog number
FGCBP2473-1
Price (MYR)
491.00
EA
Add to cart
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Price (MYR)
491.00
EA
Add to cart
Chemical Identifiers
CAS
77-86-1, 6381-92-6, 7647-01-0, 7732-18-5
Specifications
Chemical Name or Material
Tris-EDTA
Name Note
1X Solution, pH 8.0
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Color
Colorless
View more
Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.