Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.), Fisher BioReagents™
Have Questions?
Change view
Quantity:
500 mL
Packaging:
Poly Bottle
Catalog number FGCBP1757-500
Price (MYR)
592.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
592.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris Hydrochloride
Name Note1M Solution, pH 7.5
CAS77-86-1
ChemAlert Storage SymbolGray
ColorUndesignated
View more
Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.)
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.5 provide convenient and highly purified solutions.