Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Fisher Chemical
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max. | CAS: 9012-36-6 | C12H18O9
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Poly Bottle
Catalog number FGCBP160-500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose
Name NoteLow-EEO/Multi-Purpose
Physical FormPowder/Solid
CAS9012-36-6
ChemAlert Storage SymbolGray
View more
Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max.