Check Order Status
Microbiology Media Additives: Yeast Extract
Actual product may vary
Fisher Chemical
Microbiology Media Additives: Yeast Extract
Additive for preparing bacterial nutrient media
Have Questions?
Change view
Catalog Number
Quantity
FGCBP1422-500
500 g
FBRBP1422-100
100 g
FBRBP1422-500
500 g
FBRBP1422-2
2000 g
4 Options
Catalog number
FGCBP1422-500
Price (MYR)
651.00
EA
Add to cart
Quantity:
500 g
Price (MYR)
651.00
EA
Add to cart
Water soluble portion of autolyzed fresh yeast used in bacteriological culture media
Specifications
Description
Bacterial Culture
Form
Dehydrated
Grade
Molecular genetics
Media Type
Yeast Extract
Product Type
Microbiology Media
Packaging Type
Poly Bottle
Quantity
500 g