Check Order Status
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Element Search
Structure Search
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical™
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid
Have Questions?
Catalog Number
Quantity
FGCB476577
also known as FGCB/4765/77
50 Tablets
Catalog number
FGCB476577
also known as FGCB/4765/77
Price (MYR)
350.00
EA
Add to cart
Quantity:
50 Tablets
Request bulk or custom format
Price (MYR)
350.00
EA
Add to cart
Specifications
Color
White
pH
7
Physical Form
Solid
Quantity
50 Tablets
Formulation
Disodium Orthophosphate Heptahydrate, Sodium Chloride, Dipotassium Phosphate
View more
Buffer Tablets, pH 7 (Phosphate), for pH measurement, to make-up volume of 100ml, Fisher Chemical, White, Packaging: Glass Jar, Quantity: 50 tablets, MDL Number: 81857, Physical Form: Solid