▸
Acids
▸
Inorganic Acids
▸
Hydrofluoric Acid
▸
Hydrofluoric Acid 47-51%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Hydrofluoric Acid 47-51%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCA513-P500
500 mL
Catalog number
FGCA513-P500
Price (MYR)
1,278.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,278.00
Each
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7664-39-3
Melting Point
-35°C
Boiling Point
105°C
View more