Check Order Status
Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical
Element Search
Structure Search
Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical
Have Questions?
Catalog number
FGCA3320PB17
also known as FGCA/3320/PB17
Price (MYR)
342.00
Price (MYR)
342.00
EA
Add to cart
Specifications
pH
12
Color
Colorless
Boiling Point
38°C
CAS
1336-21-6
Melting Point
-57°C
View more
2.5L, Plastic Coated Glass bottle