Check Order Status
Acetone, for Electronic Use (MOS), Fisher Chemical™
Element Search
Structure Search
Acetone, for Electronic Use (MOS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCA060121
also known as FGCA/0601/21
5 L
Aluminum can
Catalog number
FGCA060121
also known as FGCA/0601/21
Price (MYR)
516.00
EA
Add to cart
Quantity:
5 L
Packaging:
Aluminum can
Request bulk or custom format
Price (MYR)
516.00
EA
Add to cart
Chemical Identifiers
CAS
67-64-1
Specifications
Packaging
Aluminum can
CAS
67-64-1
Vapor Pressure
247mbar at 20°C
Viscosity
0.32 mPaS at 20°C
Vapor Density
2
View more