Check Order Status
GP™ MAXX Enhanced Universal Absorbent Pads
Actual product may vary
GP™ MAXX Enhanced Universal Absorbent Pads
General purpose absorbent for all fluids (water, petroleum and chemical-based)
Have Questions?
Catalog Number
Capacity (English)
Packaging Quantity
Weight Capacity
FDMGP100
30 gal.
100/Cs.
Heavyweight
Catalog number
FDMGP100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (English):
30 gal.
Packaging Quantity:
100/Cs.
Weight Capacity:
Heavyweight
Single-ply construction
Class: Universal
Available in various weights, (Heavy, Medium and Light)
Made with MAXX: greater performance without the extra weight
Specifications
Certifications/Compliance
Pass 2011/65/EU
Color
Gray
Capacity (English)
30 gal.
Capacity (Metric)
30 gal./Cs.
Material
Polypropylene
Type
Absorbent Pad
For Use With (Application)
Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge Ponds
Packaging Quantity
100/Cs.
Weight Capacity
Heavyweight
Dimensions (L x W)
38 x 48.26 cm (15 x 19 in.)