Check Order Status
Hydrogen Peroxide, 6%
Element Search
Structure Search
Hydrogen Peroxide, 6%
Have Questions?
Catalog Number
Concentration
FCSS25361
6% Solution - Stabilized
Catalog number
FCSS25361
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Concentration:
6% Solution - Stabilized
Request bulk or custom format
Specifications
Chemical Name or Material
Hydrogen Peroxide
Shelf Life
1 year
Color
Colorless
CAS
7722-81-1
CAS
7732-18-5
View more