Check Order Status
Sodium Thiosulfate, 0.0394 N, for Iodine Adsorption Number of Carbon Black, Ricca Chemical
Element Search
Structure Search
Sodium Thiosulfate, 0.0394 N, for Iodine Adsorption Number of Carbon Black, Ricca Chemical
For Iodine Adsorption Number of Carbon Black Testing
Have Questions?
Catalog Number
Quantity
FCS793032
1 L
Catalog number
FCS793032
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS
10102-17-7, 7732-18-5, 497-19-8
Specifications
Chemical Name or Material
Sodium Thiosulfate
Color
Colorless
CAS
10102-17-7
CAS Min %
0.77
CAS Max %
0.8
View more