Check Order Status
Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical
Element Search
Structure Search
Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical
5400 ppm TDS as NaCl
Have Questions?
Catalog Number
Quantity
Packaging
FCS2246-16
500 mL
Natural Poly Bottle
Catalog number
FCS2246-16
Price (MYR)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7732-18-5
Specifications
Chemical Name or Material
Conductivity/TDS Standard
Name Note
10mS/cm ((10, 000μmho/cm) at 25°C, 5400ppm as NaCl
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS Min %
97.0
View more