Chart Recorders for Ultra-Low Temperature Freezers
Chart Recorders for Ultra-Low Temperature Freezers
Actual product may vary
Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FCR500LERevco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only)Inkless Chart Recorder, Field Installed
17020
also known as 17020TF
HERAfreeze HFU-B, Forma 900, Revco ExF/DxF, TSE/TSD SeriesInk Chart Paper (pack of 50)
201240TSE/TSD Series, Forma 900, HERAfreeze HFU-B, Revco ExF/DxFInk Chart Recorder, Field Installed
201241TSE/TSD Series, Forma 900, HERAfreeze HFU-B, Revco ExF/DxFInk Chart Recorder, Factory Installed
201248HERAfreeze HFU-B, Forma 900, Revco ExF/DxF, TSE/TSD SeriesInkless Chart Recorder, Field Installed
201249Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD SeriesInkless Chart Recorder, Factory Installed
245231HERAfreeze HFU-B, Forma 900, Revco ExF/DxF, TSE/TSD SeriesChart Recorder Pen, Red
245232Forma 900, HERAfreeze HFU-B, Revco ExF/DxF, TSE/TSD SeriesChart Recorder Pen, Blue
6383-6WForma 88300, HERAfreeze HFU300T, Revco UxF300Inkless Chart Recorder, Field Installed
Catalog number FCR500LE
Price (MYR)
14,766.00
EA
Add to cart
For Use With (Equipment):
Revco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only)
Type:
Inkless Chart Recorder, Field Installed
Price (MYR)
14,766.00
EA
Add to cart
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Features:
  • Seven-day recorder
  • Functions to -150°C
  • Field or factory installed
Ordering information:
  • Field-installed units must be installed by a qualified professional
Specifications
TypeInkless Chart Recorder, Field Installed
For Use With (Equipment)Revco RDE, Forma FDE, TDE Series, HDE Series (500 box capacity models only)
Monitoring OptionsRecorder
HoldsBoxes (500)
Unit SizeEA