Check Order Status
Urease (Powder)
Urease (Powder)
Have Questions?
Catalog Number
Quantity
FCMU21-25
25 g
Catalog number
FCMU21-25
Price (MYR)
-
Quantity:
25 g
Specifications
Color
White to Yellow
Quantity
25 g
Product Type
Urease
Form
Solid
Unit Size
EA
Contents & Storage
-20°C