Check Order Status
Potassium Iodide Solution (10% W/v), Fisher Chemical™
Element Search
Structure Search
Potassium Iodide Solution (10% W/v), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMSP242-1
1 L
Glass Bottle
Catalog number
FCMSP242-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Iodide Solution
Color
Undesignated
Name Note
10% (w/v)
CAS
7681-11-0
ChemAlert Storage Symbol
Gray
View more