Check Order Status
o-Tolidine Dihydrochloride Solution (0.1% w/v), Fisher Chemical™
Actual product may vary
o-Tolidine Dihydrochloride Solution (0.1% w/v), Fisher Chemical™
For determination of chlorine in water
Have Questions?
Catalog Number
Quantity
Packaging
FCMSO14-500
500 mL
Glass Bottle
Catalog number
FCMSO14-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Specifications
Color
Clear
pH Range
1
Format
Liquid
Packaging
Glass Bottle
Quantity
500 mL
Type
o-Tolidine Dihydrochloride Solution