Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Fisher Chemical
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FCMSB115-500500 mLPoly Bottle
Catalog number FCMSB115-500
Price (MYR)
369.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
369.00
EA
Add to cart
Chemical Identifiers
CAS6381-92-6, 584-08-7, 1332-77-0, 1310-58-3
Specifications
AbsorbancePass Test
Chemical Name or MaterialBuffer Solution, pH 10.00, Color-Coded Blue
ChemAlert Storage SymbolGray
Melting Point0°C
Boiling Point100°C
View more