Check Order Status
Safranin O (Certified Biological Stain), Fisher Chemical™
Element Search
Structure Search
Safranin O (Certified Biological Stain), Fisher Chemical™
For use in histology, cytology, bacteriology (staining)
Have Questions?
Catalog Number
Quantity
Packaging
FCMS670-25
25 g
Glass Bottle
Catalog number
FCMS670-25
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Safranin O
CAS
477-73-6
ChemAlert Storage Symbol
Gray
Bio. Stain Comm. Certification
Pass Test
Dye Content
Report %
View more