Ruthenium Solution for ICP, 10g/L, HCl Matrix, Fisher Chemical™
Product Image
Fisher Chemical
Ruthenium Solution for ICP, 10g/L, HCl Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Have Questions?
Catalog NumberQuantityPackaging
FCMS248-500100 mLBottle
Catalog number FCMS248-500
Price (MYR)
503.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Bottle
Request bulk or custom format
Price (MYR)
503.00
EA
Add to cart
Specifications
Shelf Life24 months (12 months after opening)
PackagingBottle
Health Hazard 1Danger
Health Hazard 2H225; H304; H315; H336; H410
Health Hazard 3P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
View more