Check Order Status
Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog number
FCMS162-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
112926-00-8, 7646-79-9
Specifications
Chemical Name or Material
Silica Gel Desiccant, Indicating
Color
Blue
Physical Form
Solid
Name Note
4-10 Mesh, Grade 48
CAS
112926-00-8
View more