Check Order Status
Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMP250-500
500 g
Poly Bottle
Catalog number
FCMP250-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Hydroxide
pH
13.5
Color
Yellow
Boiling Point
1320°C
CAS
1310-58-3
View more