Check Order Status
Hydrogen Peroxide Solution 30-32%, Optima™, for Ultra Trace Analysis, Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Hydrogen Peroxide Solution 30-32%, Optima™, for Ultra Trace Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FCMP170-500
500 mL
Catalog number
FCMP170-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Color
Undesignated
CAS
7722-84-1
Boiling Point
108°C
Melting Point
-33°C
Molecular Formula
H2O2
View more