Check Order Status
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
Element Search
Structure Search
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
High purity solvent for GC and pesticide residue analysis
Have Questions?
Change view
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Catalog number
FCME120-4
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Specifications
Chemical Name or Material
Petroleum Ether
Cleanup Symbol
Solvent
CAS
8032-32-4
ChemAlert Storage Symbol
Red
Color
Colorless
View more
Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
Also meets ACS specifications
Intended for research use only