Check Order Status
Dextrose Anhydrous (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical™
Element Search
Structure Search
Dextrose Anhydrous (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMD19-212
2.5 kg
Flint Glass
Catalog number
FCMD19-212
Price (MYR)
686.00
EA
Add to cart
Quantity:
2.5 kg
Packaging:
Flint Glass
Request bulk or custom format
Price (MYR)
686.00
EA
Add to cart
Specifications
Chemical Name or Material
Dextrose Anhydrous
CAS
50-99-7
ChemAlert Storage Symbol
Gray
Melting Point
146.1°C
pH
5.9
View more