Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
Fisher Chemical
Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
1LT Dichloromethane, for residue analysis, Distol™, stabilised with amylene
Have Questions?
Catalog NumberQuantityPackaging
FCMD151-44 LAmber Soda Lime Glass
Catalog number FCMD151-4
Price (MYR)
844.00
Bottle
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (MYR)
844.00
Bottle
Specifications
Chemical Name or MaterialMethylene Chloride
Cleanup SymbolSolvent
Melting Point-97°C
Absorbance1 AU max. at 233nm, 0.12 AU max. at 240nm, 0.005 AU max. at 254nm
CAS75-09-2
View more
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
  • Contains approx. 15- 200 ppm amylene as preservative
  • Also meets ACS specifications
  • Intended for research use only