Check Order Status
Dimethyl Sulfoxide (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Dimethyl Sulfoxide (Certified ACS), Fisher Chemical™
Note: Sold for nonmedical use only. Not to be used or sold as a drug for humans or animals
Have Questions?
Catalog Number
Quantity
Packaging
FCMD128-1
1 L
HDPE Bottle
Catalog number
FCMD128-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
HDPE Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Dimethyl Sulfoxide
Melting Point
18.4°C
CAS
67-68-5
ChemAlert Storage Symbol
Gray
Acidity
Pass Test (USP)
View more