Check Order Status
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™
Element Search
Structure Search
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™
DNase-, RNase- and Protease-Free. | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Catalog number
FCMBP152-500
Price (MYR)
540.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
540.00
EA
Add to cart
Specifications
Absorbance
0.05 AU max. at 280nm
Chemical Name or Material
Tris Base
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Melting Point
168.5°C
View more
DNase-, RNase- and Protease-Free.
Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.