Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
HPLC mobile phase for analytical and preparative separations
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Packaging:
View all
Wide Neck Amber Bottle
Catalog number FCMA998-1
Price (MYR)
628.00
EA
Add to cart
Quantity:
1 L
Packaging:
Wide Neck Amber Bottle
Request bulk or custom format
Price (MYR)
628.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile
Cleanup SymbolSolvent
Melting Point-46°C
Absorbance0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
CAS75-05-8
View more