Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical
Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical
Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical
Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical
Fisher Chemical
Sulfuric acid 93-98%, Trace Metal™, for trace metal analysis, Fisher Chemical
Have Questions?
Catalog number FCMA510-P212
Price (MYR)
1,475.00
Price (MYR)
1,475.00
EA
Add to cart
Specifications
pH1
ColorColorless
CAS7664-93-9
Melting Point10°C
Boiling Point290°C
View more