Sulfuric Acid (Optima™), Fisher Chemical™
Sulfuric Acid (Optima™), Fisher Chemical™
Sulfuric Acid (Optima™), Fisher Chemical™
Fisher Chemical
Sulfuric Acid (Optima™), Fisher Chemical™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
FEP Bottle
Catalog number FCMA468-1
Price (MYR)
9,159.00
Each
Quantity:
1 L
Packaging:
FEP Bottle
Request bulk or custom format
Price (MYR)
9,159.00
Each
Specifications
Chemical Name or MaterialSulfuric Acid
pH0.3
Cleanup SymbolAcid
ColorColorless to Brown
CAS7664-93-9
View more
  • Certificate of Lot Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintainproduct integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture available on COA.