HPLC mobile phase for analytical and preparative separations
Have Questions?
Catalog Number
Quantity
Packaging
FCMA451-4
4 L
Glass Bottle
Catalog number FCMA451-4
Price (MYR)
586.00
BOT
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
586.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Propanol
pH7
Cleanup SymbolSolvent
Melting Point-89.5°C
Absorbance0.025 AU max. at 254nm, 0.13 AU max. at 230nm, 0.25 AU max. at 220nm, 1.00 AU max. at 205nm
View more
Safe-Cote™ bottles that provide the purity of glass and most of the benefits for storing and dispensing chemicals. If they break, glass fragments and liquids are more likely to remain trapped.