Check Order Status
Phosphorus Pentoxide (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Phosphorus Pentoxide (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA245-500
500 g
Glass Bottle
Catalog number
FCMA245-500
Price (MYR)
624.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
624.00
EA
Add to cart
Specifications
Boiling Point
360°C
Chemical Name or Material
Phosphorus Pentoxide
Melting Point
340°C
CAS
1314-56-3
ChemAlert Storage Symbol