Check Order Status
Acetonitrile (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetonitrile (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Have Questions?
Change view
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Catalog number
FCMA21-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Acetonitrile
Cleanup Symbol
Solvent
Melting Point
-46°C
CAS
75-05-8
ChemAlert Storage Symbol
Red
View more