Check Order Status
Acetone (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetone (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Have Questions?
Catalog Number
Quantity
Packaging
FCMA18-4
4 L
Glass Bottle
Catalog number
FCMA18-4
Price (MYR)
523.00
BOT
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
523.00
BOT
Add to cart
Specifications
Chemical Name or Material
Acetone
pH
7
Cleanup Symbol
Solvent
Melting Point
-95°C
CAS
67-64-1
View more