Check Order Status
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number
FCMA144-500
Price (MYR)
570.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
570.00
EA
Add to cart
Specifications
pH
<1
Cleanup Symbol
Acid
Color
Colorless
CAS
7647-01-0
Melting Point
-35°C
View more