Check Order Status
Hydrochloric Acid (Technical), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid (Technical), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA142-212
2.5 L
Catalog number
FCMA142-212
Price (MYR)
541.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
541.00
EA
Add to cart
Specifications
Appearance
Liquid
Chemical Name or Material
Hydrochloric Acid
Cleanup Symbol
Acid
CAS
7647-01-0
ChemAlert Storage Symbol