AREX 5 Hotplate Stirrer
AREX 5 Hotplate Stirrer
Actual product may vary
AREX 5 Hotplate Stirrer
Versatile AREX 5 Hotplate Stirrer with CerAlTop™ plate is upgradable with the VTF digital thermoregulator for direct liquid temperature control.
Have Questions?
Catalog NumberPlug Type
FBV17872017EU/UK
Catalog number FBV17872017
Price (MYR)
2,931.00
EA
Add to cart
Plug Type:
EU/UK
Price (MYR)
2,931.00
EA
Add to cart
UPGRADE TO DIGITAL WITH VTF FOR A PRECISE THERMOREGULATION
  • Can work stand-alone or be upgraded to digital by connecting the VTF digital temperature controller
  • The 135 mm diameter aluminium alloy top plate ensures great temperature homogeneity and optimum heat transfer across the entire surface
EASY-TO-USE AND SAFE
  • Two illuminated indicators always inform the operator when the heating is on and the temperature is above 50°C
  • Insulating disk to protect internal parts
  • The 135 mm diameter top is protected from chemicals and scratches by the superior resistant CerAlTop™ ceramic coating
  • IP42 certified
POWERFUL STIRRING PERFORMANCE FOR ALL LAB APPLICATIONS
  • Powerful Brushless motor ensuring power and resistance over time and stirs volumes up to 20 liters (H2O)
  • Stirring speed from 100 up to 1500 rpm
  • SpeedServo™ torque compensation technology that ensures constant speed even when the viscosity of the medium changes
Specifications
Top Plate MaterialCeramic-coated Aluminum Alloy
Voltage230 V
Safety FeaturesIlluminated Icon (when the temperature is above 50°C)
Body MaterialEpoxy-painted Aluminum
Controller TypeAnalog
Size135 mm dia.
Temperature (Metric)Ambient to 310°C
Temperature (English)Ambient to 590°F
Frequency50/60 Hz
Wattage630 W
Weight (English)3.75 lb.
Weight (Metric)1.7 kg
TypeStirring Hotplate
ShapeRound
Max. Sample Load20 L
Plug TypeEU/UK
No. of Positions1
Max. Container Size20 L
Stirring Range100 to 1500 rpm
Warranty3 Years
Dimensions (LxWxH)160 x 85 x 270 mm
DisplayAnalog
Unit SizeEA