Check Order Status
Tetracycline Hydrochloride, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Tetracycline Hydrochloride, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP912-100
100 g
Catalog number
FBRBP912-100
Price (MYR)
657.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
657.00
BOT
Add to cart
Specifications
Chemical Name or Material
Tetracycline Hydrochloride
Type
Tetracycline Hydrochloride
Physical Form
Solid
CAS
64-75-5
ChemAlert Storage Symbol
Gray
View more
Tetracycline blocks protein synthesis and is used to select tetracycline-resistant recombinant plasmids.