Check Order Status
PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents
Actual product may vary
Fisher Chemical
PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents
Have Questions?
Change view
Catalog Number
Quantity
FBRBP661-50
50 L
FBRBP661-10
10 L
2 Options
Catalog number
FBRBP661-50
Price (MYR)
1,012.00
EA
Add to cart
Quantity:
50 L
Price (MYR)
1,012.00
EA
Add to cart
This product provides a convenient way to make large volumes of standard 1X Phosphate Buffer; each bottle contains 98.9g or 492g for reconstitution in 10L or 50L of water, respectively.
Specifications
Color
White
Recommended Storage
Room temperature
Physical Form
Powder
Quantity
50 L
Packaging
Poly Bottle